Micro-Cut Kevorkian-Younge Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8905
10199-188EA 1554.49 USD
10199-188
Micro-Cut Kevorkian-Younge Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Micro-Cut Kevorkian-Younge Biopsy Punch is a non-ratcheted, finger ring instrument used for obtaining tissue samples in gynecological procedures
The Micro-Cut instrument features a knife honed edge on the upper jaw for a cleaner and more precise cut of the tissue. The shanks are short and angle at a box lock to a long, narrow shaft that is 10 inches long. The tip has a 3mm x 9.7mm rectangular bite with 4 lower teeth to grasp and excise the tissue for biopsy.