Mini Tischler Universal Style Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-7553
10198-882EA 1581.84 USD
10198-882
Mini Tischler Universal Style Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Mini Tischler Universal Style Biopsy Punch Forceps are a non-ratcheted, double spring OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy
The Mini Tischler bite, 2.3x4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. It also features a single, lower tooth used to grasp difficult to reach tissue. This product is a complete instrument which consists of a handle and a detachable cannula/tip, so other universal tip styles may be used with one handle. The length of the cannula/tip is approximately 20.3cm (8").