Oval Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-7525
10198-876EA 1564.8 USD
10198-876
Oval Biopsy Punch, OR Grade, Sklar
Biopsy Punches
This Oval Biopsy Punch Forceps are a locking, double-spring plier instrument used for obtaining uterine tissue for biopsy.
  • Made of stainless steel
  • Censitrac Ready products are industry standard
  • Reusable
  • Latex-free
The locking wheel at the base of the shaft has a smooth, beveled side with the rest being cross-serrated, which makes the lock easy to engage. This locks the jaws and the tissue sample in place while the instrument is removed from the cervical canal, thus reducing the risk of losing the tissue sample as the instrument is withdrawn.
The plier handle features a horn for thumb placement as well as a hook and two finger rests to provide maximum comfort and greater control. The bite of this instrument is the unique Burke-style punch and produces a 3x5mm oval tissue sample.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.