Weingartner Ear Forceps, Sklar
Supplier: Sklar
67-8155
10198-480EA 766.93 USD
10198-480
Weingartner Ear Forceps, Sklar
Forceps
Alligator style forceps with serrated jaw to minimize slippage. Useful in middle ear procedures where the powerful grasp and curved jaws assist in the removal of foreign objects.
  • Finger ring handles
  • Non locking
  • Reusable
  • Latex-free