Hartmann Ear Speculum, OR Grade, Sklar
Supplier: Sklar
67-6004
10198-408EA 52.85 USD
10198-408
Hartmann Ear Speculum, OR Grade, Sklar
Specula
The Sklar Hartman Ear Speculum is a cone/funnel-shaped attachment for an otoscope that allows examination of the ear canal and eardrum
The speculum is inserted into the ear canal and the otoscope handle provides a light source and low-power magnification so the practitioner can view the eardrum. Disease and infections of the middle or inner ear can largely be detected by this examination. This device is a round attachment with a 4mm diameter.