OxyPouch™ Oxygen Barrier Bag, Oxyrase®
Supplier: Oxyrase Inc
A-OP-0100
101975-878PK 85.17 USD
101975-878
OxyPouch™ Oxygen Barrier Bag, Oxyrase®
Anaerobic Systems
The OxyPouch™ is a foil oxygen barrier bag, designed and specifically sized to hold OxyPlates (or other type petri dishes) and help prevent oxidation of plates during incubation and or storage.
The OxyPouch™ is designed and constructed of materials to ensure that once the bag has been properly sealed with a heat sealer, oxygen cannot permeate through the bag. This prevents any further oxidation of the plate sealed within the bag.
Each OxyPouch™ also has one clear side with which to easily read the labeled portion of the base plate, without the need to open the bag. The upper portion of the bag is perforated, to make opening simple, without the need for scissors.
Ordering information: OxyPouch™ is shipped direct to customers.