Actinomycin D ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT955-250UCI
101974-024EA 2576.15 USD
101974-024
Actinomycin D ≥97% (by HPLC), MORpure™
Actinomycin D
Formula: C₆₂H₈₆N₁₂O₁₆
MW: 1255.5 g/mol
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 2910
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR