Puromycin dihydrochloride, [8-3H(N)] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT785-250UCI
101973-614EA 2576.15 USD
101973-614
Puromycin dihydrochloride, [8-3H(N)] ≥97% (by HPLC), MORpure™
Puromycin dihydrochloride, [8-3H(N)]
CAS Number: 73430-35-0
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR