2'-Deoxycytidine-5'-monophosphate, diammonium salt, [5-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT698-250UCI MT698-1MCI
101973-428EA 2700.38 USD
101973-428 101973-430
2'-Deoxycytidine-5'-monophosphate, diammonium salt, [5-3H] ≥96%
2'-Deoxycytidine-5'-monophosphate, diammonium salt, [5-3H]
CAS Number: 339340-57-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR