2'-Deoxyuridine-5'-monophosphate, diammonium salt, [5-3H] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MT606-250UCI MT606-1MCI MT606-5MCI
101973-186EA 2700.38 USD
101973-186 101973-188 101973-190
2'-Deoxyuridine-5'-monophosphate, diammonium salt, [5-3H] ≥96%
2'-Deoxyuridine-5'-monophosphate, diammonium salt, [5-3H]
CAS Number: 339163-05-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR