Isoeugenol

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Propenylguaiacol, 4-Hydroxy-3-methoxy-1-propenylbenzene, 2-Methoxy-4-(prop-1-en-1-yl)phenol

MT1962-50UCI
101972-880EA 1601.92 USD
101972-880
Isoeugenol
Isoeugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00009285
CAS Number: 97-54-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR