Vitamin B12 (cyanocobalamin)

Supplier: MORAVEK BIOCHEMICALS MS

MT1953-50UCI
101972-854EA 1601.92 USD
101972-854
Vitamin B12 (cyanocobalamin)
Vitamin B12 (cyanocobalamin)
Formula: C₆₃H₈₈CoN₁₄O₁₄P
MDL Number: MFCD00151092
CAS Number: 68-19-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR