Nifurtimox, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 3-Methyl-4-(((5-nitrofuran-2-yl)methylene)amino)thiomorpholine 1,1-dioxide, Nifurtimox

MT1877-250UCI MT1877-1MCI
101972-678EA 2700.38 USD
101972-678 101972-680
Nifurtimox, [3H] ≥97%, MORpure™
Nifurtimox, [3H]
Formula: C₁₀H₁₃N₃O₅S
MW: 287.290 g/mol
MDL Number: MFCD00869254
CAS Number: 23256-30-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR