Estriol, [2,4-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1785-250UCI
101972-618EA 2700.38 USD
101972-618
Estriol, [2,4-3H] ≥97%, MORpure™
Estriol, [2,4-3H]
CAS Number: 19590-79-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR