L-2-Aminobutyric acid, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1776-250UCI
101972-598EA 2700.38 USD
101972-598
L-2-Aminobutyric acid, [3H] ≥97%, MORpure™
L-2-Aminobutyric acid, [3H]
CAS Number: 82144-85-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR