2-Methyl-D,L-aspartic acid, disodium salt, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1747-250UCI
101972-546EA 2700.38 USD
101972-546
2-Methyl-D,L-aspartic acid, disodium salt, [3H] ≥97%, MORpure™
2-Methyl-D,L-aspartic acid, disodium salt, [3H]
CAS Number: 57282-56-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR