Succinic acid, [1,4-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC238-50UCI MC238-250UCI MC238-1MCI
101971-508EA 765 USD
101971-508 101971-510 101971-512
Succinic acid, [1,4-14C] ≥98%, MORpure™
Succinic acid, [1,4-14C]
CAS Number: 13613-74-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR