D-Phenylalanylamide, [ring-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2305-50UCI
101971-376EA 5387.69 USD
101971-376
D-Phenylalanylamide, [ring-14C(U)] ≥98%, MORpure™
D-Phenylalanylamide, [ring-14C(U)]
CAS Number: 2088576-48-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR