Chinomethionat, [quinoxaline-2,3-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2294-50UCI
101971-352EA 5387.69 USD
101971-352
Chinomethionat, [quinoxaline-2,3-14C] ≥98%, MORpure™
Chinomethionat, [quinoxaline-2,3-14C]
CAS Number: 2088204-27-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR