Forchlorfenuron, [pyridine-2,6-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2247-50UCI
101971-260EA 5387.69 USD
101971-260
Forchlorfenuron, [pyridine-2,6-14C] ≥98% (by HPLC), MORpure™
Forchlorfenuron, [pyridine-2,6-14C]
CAS Number: 2088573-52-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR