D-Phenylalanine, [ring-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2239-50UCI MC2239-250UCI
101971-250EA 5387.69 USD
101971-250 101971-252
D-Phenylalanine, [ring-14C(U)] ≥98%, MORpure™
D-Phenylalanine, [ring-14C(U)]
CAS Number: 2088617-18-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR