(S)-Amlodipine, malic acid salt, [dihydropyridine-4-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2236-10UCI
101971-242EA 1621.54 USD
101971-242
(S)-Amlodipine, malic acid salt, [dihydropyridine-4-14C] ≥98% (by HPLC), MORpure™
(S)-Amlodipine, malic acid salt, [dihydropyridine-4-14C]
CAS Number: 2088575-59-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR