Stearoyl coenzyme A

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Stearoyl Coenzyme A, Free Acid

MC1439-10UCI MC1439-50UCI
101970-564EA 2040 USD
101970-564 101970-566
Stearoyl coenzyme A
Stearoyl coenzyme A
Formula: C₃₉H₇₀N₇O₁₇P₃S
MW: 1034.002 g/mol
MDL Number: MFCD00079568
CAS Number: 362-66-3
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR