D-Arginine, [1-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC1402-50UCI MC1402-250UCI
101970-492EA 5387.69 USD
101970-492 101970-494
D-Arginine, [1-14C] ≥97%
D-Arginine, [1-14C]
CAS Number: 289623-53-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR