Bromoacetic acid, [2-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1257-50UCI MC1257-250UCI MC1257-1MCI
101970-270EA 5387.69 USD
101970-270 101970-272 101970-274
Bromoacetic acid, [2-14C] ≥98%, MORpure™
Bromoacetic acid, [2-14C]
CAS Number: 50718-91-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR