3'-Azido-3'-deoxythymidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT

M817-25MG
101970-006EA 778.08 USD
101970-006
3'-Azido-3'-deoxythymidine
3'-Azido-3'-deoxythymidine
Formula: C₁₀H₁₃N₅O₄
MW: 267.25 g/mol
MDL Number: MFCD00006536
CAS Number: 30516-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR