Sklar Black™ Collin Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3946
10197-948EA 484.03 USD
10197-948
Sklar Black™ Collin Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ Collin Vaginal Speculum is a gynecological instrument.
  • Used for performing pelvic examinations
  • Pap Smears
  • Cervix exams
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. Unlike many other vaginal specula, there is no handle with the Collin device and the blades open laterally/horizontally versus vertically.
The practitioner can best determine which type and size speculum is most appropriate for the patient. This product features a permanent smoke evacuation tube (not shown).
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.