Collin Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3945
10197-946EA 312.59 USD
10197-946
Collin Vaginal Speculum, OR Grade, Sklar
Specula
Sklar's® Collin Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
  • Double blade duckbill speculum inserted with the blades closed
  • Industry standard
Unlike many other vaginal specula, there is no handle with the Collin device and the blades open laterally/horizontally versus vertically.