Kratz-Barraquer Wire Speculum, OR Grade, Sklar
Supplier: Sklar
65-1754
10197-918EA 84.96 USD
10197-918
Kratz-Barraquer Wire Speculum, OR Grade, Sklar
Specula
The Kratz-Barraquer Wire Speculum is an ophthalmic instrument used to retain the eyelids open, often during cataract or phacoemulsification procedures
  • Light weight
It is utilized to provide the surgeon with increased visualization and access to the surgical site. To a degree, the surgeon can increase the tension of the instrument by manipulating the wire where the speculum is held, as it is slightly maleable. In situations where a wire speculum will not provide enough force to open the lids sufficiently, a speculum with a screw mechanism may be used.