Lawrence Forceps, OR Grade, Sklar
Supplier: Sklar
55-2685
10197-570EA 319.84 USD
10197-570
Lawrence Forceps, OR Grade, Sklar
Forceps
Lawrence Forceps are a ratcheted, finger ring instrument used for controlling blood flow is vessels, often in cardiovascular and thoracic procedures.
The long shanks allow this instrument to be used in difficult to reach areas, while the curved, serrated jaws can clamp obscured vessels.