Castroviejo Lachrymal Dilator, OR Grade, Sklar®
Supplier: Sklar
65-6960
10197-422EA 87.73 USD
10197-422
Castroviejo Lachrymal Dilator, OR Grade, Sklar®
Dilators
This is a double-ended instrument most commonly used in ophthalmologic, lacrimal system disorder procedures, and oculofacial plastic surgery.
  • Straight and round
  • Two tapered tips
The lachrymal dilator may be used to open the tear duct to increase tear flow, access the lachrymal system for obstruction removal, and repair damage to or disease of the lachrymal system.