Barraquer Cyclodialysis Spatula, OR Grade, Sklar
Supplier: Sklar
65-4068
10197-380EA 155.77 USD
10197-380
Barraquer Cyclodialysis Spatula, OR Grade, Sklar
Spatulas
Sklar's Barraquer Cyclodialysis Spatula is an ophthalmic instrument primarily used for repositioning the iris.
Sklar offers this product with a 15 mm angled blade, blunt 0.5 mm tip, and a length of 4³/₄ inches.