Graves Wide View Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3705
10197-208EA 191.65 USD
10197-208
Graves Wide View Vaginal Speculum, OR Grade, Sklar
Specula
Graves wide view vaginal speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
It is a standard double blade duckbill speculum that features a wide proximal opening for instrumentation. It is inserted with the blades closed. The blades are then opened by an external lever and screw.