Henrotin Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3555
10197-196EA 1266.47 USD
10197-196
Henrotin Weighted Vaginal Speculum, OR Grade, Sklar
Specula
Sklar's Henrotin Weighted Vaginal Speculum utilizes a 3 lbs weight to hold the narrow, angled blade in place for use in OB/GYN procedures. The blade is angle downward 30® to provide the user with a greater view of the vaginal canal.