β-L-Cytidine ≥98%, MORpure™
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
4-Amino-1-((2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, L-Cytidine
M320-1MG
M320-5MG
101969-888EA
294.23
USD
101969-888
101969-890
β-L-Cytidine ≥98%, MORpure™
β-L-Cytidine
Formula:
C₉H₁₃N₃O₅ MW: 243.2166 g/mol |
MDL Number:
MFCD02683605 CAS Number: 26524-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...