L-Phenylalanine

Supplier: MORAVEK BIOCHEMICALS MS

M1888-1MG M1888-5MG
101969-814EA 294.23 USD
101969-814 101969-816
L-Phenylalanine
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR