β-L-Inosine ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: L-Inosine, 9-((2S,3S,4R,5S)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

M1807-1MG M1807-5MG
101969-668EA 294.23 USD
101969-668 101969-670
β-L-Inosine ≥98%, MORpure™
β-L-Inosine
Formula: C₁₀H₁₂N₄O₅
MW: 268.226 g/mol
MDL Number: MFCD00673221
CAS Number: 21138-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR