Valacyclovir hydrochloride hydrate
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride
M1571-1MG
M1571-5MG
101969-260EA
294.23
USD
101969-260
101969-262
Valacyclovir hydrochloride hydrate
Valacyclovir hydrochloride hydrate
Formula:
C₁₃H₂₀N₆O₄ MW: 360.80 g/mol |
MDL Number:
MFCD01861507 CAS Number: 124832-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...