Aufricht Glabellar Rasp, OR Grade, Sklar
Supplier: Sklar
40-9840
10196-598EA 296.41 USD
10196-598
Aufricht Glabellar Rasp, OR Grade, Sklar
Raspatories
Aufricht Glabellar Raspatory is a debulking instrument used for filing rough bone edges in head and neck procedures.
This product has coarse, horizontal serrations and a slightly curved tip intended for forward cutting.