Fomon Raspatory and File, OR Grade, Sklar
Supplier: Sklar
40-9688
10196-586EA 410.74 USD
10196-586
Fomon Raspatory and File, OR Grade, Sklar
Raspatories
Fomon raspatory with four surfaces and file is designed for precise bone contouring in nasal reconstruction.
  • Stainless steel
Cross striations allow for either an up or down cutting motion.