Campion Uterine Forceps, OR Grade, Sklar
Supplier: Sklar
90-7898
10196-410EA 381.37 USD
10196-410
Campion Uterine Forceps, OR Grade, Sklar
Forceps
Campion Uterine Forceps are a non-ratcheted, finger ring OB/GYN instrument.
This grasping instrument features long shanks, as well as long jaws, seen in uterine forceps. It has a slight jaw type, beginning at half the jaw.
The 15-mm tips are a fenestrated, long, slightly cupped, oval shape.