Z-Type Hysterectomy Forceps, OR Grade, Sklar
Supplier: Sklar
90-2793 90-2794 90-2790
10196-386EA 623.95 USD
10196-386 10196-388 10199-144
Z-Type Hysterectomy Forceps, OR Grade, Sklar
Forceps
Z-Type Hysterectomy Forceps are a ratcheted, finger ring forceps used in vaginal and abdominal hysterectomy procedures.
The handles of these forceps are offset, so as not to obstruct the physician's view of the surgical site.