Van Buren Urethral Sounds, OR Grade, Sklar®
Supplier: Sklar
85-1065 85-1075 85-1008 85-1010 85-1012 85-1014 85-1016 85-1018 85-1028 85-1030 85-1032 85-1034 85-1036 85-1038 85-1040 85-1070 85-1020 85-1022 85-1024 85-1026
10193-324PK 1931.24 USD
10193-324 10193-386 10196-364 10196-366 10196-368 10196-370 10196-372 10196-374 10196-376 10196-378 10196-380 10196-382 10198-810 10198-812 10198-814 10198-816 10198-898 10198-900 10198-902 10198-904
Van Buren Urethral Sounds, OR Grade, Sklar®
Dilators
Sklar's® Van Buren urethral curved sound is used for dilation of the urethra to look for obstructions.
  • Made of stainless steel
  • Industry standard
  • Reusable and latex-free
  • Non sterile
  • Premium OR-grade
It is available in a variety of diameters for a range of cases.