Improved Rudd Clinic Hemorrhoid Ligator, OR Grade, Sklar
Supplier: Sklar
80-1918
10196-312EA 465.37 USD
10196-312
Improved Rudd Clinic Hemorrhoid Ligator, OR Grade, Sklar
Forceps
Improved Rudd Clinic Hemorrhoid Ligator feature an integrated resetting spring, which is detachable for easy cleaning.
  • Reusable and latex-free
  • Non-Sterile
  • Premium OR grade
The ligator is constructed of stainless steel and with industry standard.