Solid Bar Handle for Gigli Saw, OR Grade, Sklar
Supplier: Sklar
40-5520
10195-916PK 279.1 USD
10195-916
Solid Bar Handle for Gigli Saw, OR Grade, Sklar
Microtomes and Microtome Assemblies Microtome Blades and Handles
Sklar's Gigli T Saw Handle is used in conjunction with a Gigli Wire Saw Blade
This provides an even, smooth cut. This instrument is used for sawing bones, commonly amputations, but may also be used in neurosurgery for cutting cranial flaps. The Gigli Saw is an older, manual type saw, but is still used today by surgeons.
The twisted wire is connected to two T Saw Handles and used with a pulling, back and forth motion.