Stille-Horsley Bone Cutting Forceps, OR Grade, Sklar
Supplier: Sklar
40-3910
10195-820EA 1274.92 USD
10195-820
Stille-Horsley Bone Cutting Forceps, OR Grade, Sklar
Bone Cutters and Rongeurs
Stille-Horsley Bone Cutting Forceps are an orthopedic instrument often used to cut rib bones.
  • Double-action instrument provides greater pressure on the bones without as much effort by the user
  • Plier handle curves slightly at the joints, and then again, more strongly and in the opposite direction at the jaws
  • Double curvature allows the instrument to be used in tight areas
  • Jaws of the forceps have sharp blades that square when closed
  • Horned plier handle provides the user with thumb and finger rests for a more comfortable and controlling grip
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.