Mayo Guyon Kidney Clamp, OR Grade, Sklar®
Supplier: Sklar
33-1090
10195-700EA 457.47 USD
10195-700
Mayo Guyon Kidney Clamp, OR Grade, Sklar®
Clamps General Purpose Clamps
The Sklar® Mayo-Guyon Kidney Clamp is most commonly used in urological procedures where heavy tissue or organs need to be held
The heavy serrations are across the entire length of the jaws and can grasp thicker tissue.