Square End Plier, OR Grade, Sklar
Supplier: Sklar
40-1778
10195-558EA 272.99 USD
10195-558
Square End Plier, OR Grade, Sklar
Pliers
Sklar® Square End Pliers is a reusable, stainless steel, OR grade instrument.
  • Used in cardiovascular
  • Dermatology
  • General surgery
  • Orthopedics
  • Physician
  • Podiatry
  • Veterinary health
This product is straight, measuring 5.5” in length, with a cross-serrated working edge.