Gillies Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-2662
10195-046EA 238.67 USD
10195-046
Gillies Needle Holder, OR Grade, Sklar
Forceps
Gillies Needle Holder is designed for use with delicate suturing needles for closing wounds
Censitrac Ready products are industry standard and constructed of stainless steel.
This product has an offset design with curved, cross-serrated tips with scissors incorporated to make it a versatile tool, including plastic surgery procedures. The Gillies needle holder has on offset larger ring.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.