iTaq™ Universal Probes Supermix, qPCR Mastermix, Bio-Rad
Supplier: 0000013515
1725130 1725131 1725132 1725134 1725135
101946-474EA 303.08 USD
101946-474 101946-476 101946-478 101946-480 101946-482
iTaq™ Universal Probes Supermix, qPCR Mastermix, Bio-Rad
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
iTaq™ Universal Probes Supermix is a 2x ready-to-use master mix containing all components, except primers, probes, and template, for probe-based qPCR (simplex or duplex) on any real-time PCR instrument.
  • Perform simplex or duplex assays for a wide variety of targets
  • Obtain reliable and reproducible gene expression analysis results with any fluorogenic probe
  • Use for fast or standard PCR
  • Antibody-mediated hot-start iTaq™ DNA polymerase for rapid activation
  • Blend of passive reference dyes for universal real-time instrument compatibility
  • Freeze/thaw up to 10 times
  • Standard and fast thermal cycling protocols
iTaq™ Universal Probes Supermix contains antibody-mediated hot-start iTaq™ DNA polymerase, dNTPs, MgCl2, enhancers, stabilizers, and a blend of passive reference dyes (including ROX).